These products are not currently available for ordering on ThermoFisher.com. Please continue to place orders through your existing ordering channel. As part of the transition to Thermo Fisher Scientific, these products will become available through Thermo Fisher ordering channels in the coming months.
LifeASSURE™ PDA Series Filter Capsule
Thermo Scientific™
LifeASSURE™ PDA Series Filter Capsule
LifeASSURE™ PDA Series Filter Capsules use dual asymmetric PES membranes with Advanced Pleat Technology to deliver high throughput and fast flow. They support pre‐filtration and sterilizing filtration in pharma, biological and bioprocess applications requiring low protein binding.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호모델
7100037799PDA010C01AAS1
7100037808PDA010J02AAG1
7000051527PDA020C01AAG1
7000051529PDA020J01AAG1
7000051530PDA020J02AAG1
7000051531PDA020C01AAS1
7000051532PDA020C01BAS1
7000051533PDA020C02AAS1
7000051534PDA020C02BAS1
7000051535PDA020J01AAS1
7000051536PDA020J02AAS1
7000051537PDA020J03AAS1
7100005500PDA020C01BAG1
7100011132PDA020C02AAG1
7100017245PDA020J03AAG1
7100037798PDA010C01AAG1
7100037800PDA010C01BAG1
7100037801PDA010C01BAS1
7100037802PDA010C02AAG1
7100037804PDA010C02BAG1
7100037806PDA010J01AAG1
7100037810PDA010J03AAG1
7100087472PDA020C02BAG1
카탈로그 번호 7100037799
제품 가격(KRW)
-
모델:
PDA010C01AAS1
LifeAssure™ PDA filter capsules are sterilizing-grade capsules containing 0.1 or 0.2 μm PES membranes for pharma and bioprocessing. They offer high flow rates, low protein binding, and broad chemical compatibility. Used to sterilize buffers, media, and biological solutions, they can be validated for bacterial retention to ensure safety and compliance. Durable and efficient, they deliver consistent performance and available in various sizes and end fittings.
  • Validated 0.2 and 0.1 micron rated membrane provides reliable sterilizing filtration
  • Excellent contaminant capacity, flow rates, and life resulting from robust double-asymmetric membrane layer filter construction
  • Hydrophilic asymmetric polyethersulfone membrane enables wet out in aqueous solutions for integrity testing
  • Advanced Pleat Technology construction increases the useful surface area by increasing the flow paths between media pleats
  • 100% forward flow Integrity tested prior to release
  • Both pre-sterilized and sterilizable LifeASSURE™ PDA capsules are available
사양
용도(애플리케이션)Sterile Filtration, Biopharmaceutical Manufacturing
길이(영국식 단위)2.5 in.
길이(미터법)6.35 cm
모델PDA010C01AAS1
제품라인PDA
수량4/Cs.
유형Filter Capsule
필터 유형Membrane Filter
기공 크기0.1 μm
Unit SizeEach