Search
Search

| 카탈로그 번호 | 수량 | 농도 |
|---|---|---|
| A65783BE | 200 L | 1 M |
| A65782BA | 5 L | 2 M |
| A65782BB | 20 L | 2 M |
| A65782BC | 50 L | 2 M |
| A65782BD | 100 L | 2 M |
| A65782BE | 200 L | 2 M |
| A65783BA | 5 L | 1 M |
| A65783BB | 20 L | 1 M |
| A65783BC | 50 L | 1 M |
| A65783BD | 100 L | 1 M |
Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Features
Benefits
Additional information