Search
| 카탈로그 번호 | 마개 크기 | 용도(애플리케이션) |
|---|---|---|
| K7471302024 FISHERTOP -K7471302024-EA으로도 사용됨 | 20-400 | Standard Taper Joint: 24/40 |
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.