QSP Low Retention Filtered And Non-Filtered Pipette Tips
Thermo Scientific™
QSP Low Retention Filtered And Non-Filtered Pipette Tips
Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명부피(미터법)팁 스타일필터링됨제품 사이즈
TLR070RL-QPK
TLR070RL-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR102-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 10000
TLR102-QPK
TLR102-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 1000
TLR102R-QPK
TLR102R-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended Length, ConicalNon-filteredPack of 960
TLR102RS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RS-QPK
TLR102RS-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TLR102RL-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RL-QPK
TLR102RL-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TLR102RLS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RLS-QPK
TLR102RLS-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TFLR113-20-Q20 μLBeveledFilteredCase of 4800
TFLR113-20-QPK
TFLR113-20-Q으로도 사용됨
20 μLBeveledFilteredPack of 960
TFLR113-100-Q100 μLBeveledFilteredCase of 4800
TFLR113-100-QPK
TFLR113-100-Q으로도 사용됨
100 μLBeveledFilteredPack of 960
TLR070-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 10000
TLR070-QPK
TLR070-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 1000
TLR070R-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070R-QPK
TLR070R-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR070RS-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RS-QPK
TLR070RS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR070RL-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RLS-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RLS-QPK
TLR070RLS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR106-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 10000
TLR106-QPK
TLR106-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 1000
TLR106R-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106R-QPK
TLR106R-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RS-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RS-QPK
TLR106RS-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RL-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RL-QPK
TLR106RL-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RLS-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RLS-QPK
TLR106RLS-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TFLR140-200-Q200 μLMicroPointFilteredCase of 4800
TFLR140-200-QPK
TFLR140-200-Q으로도 사용됨
200 μLMicroPointFilteredPack of 960
TFLR140-300-Q300 μLMicroPointFilteredCase of 4800
TFLR140-300-QPK
TFLR140-300-Q으로도 사용됨
300 μLMicroPointFilteredPack of 960
TLR112NXL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 10000
TLR112NXL-QPK
TLR112NXL-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 1000
TLR112NXLR-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLR-QPK
TLR112NXLR-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXLRS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLRS-QPK
TLR112NXLRS-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXLRL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLRL-QPK
TLR112NXLRL-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXRLS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXRLS-QPK
TLR112NXRLS-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TFLR1121000-Q1000 μLExtended LengthFilteredCase of 4800
TFLR1121000-QPK
TFLR1121000-Q으로도 사용됨
1000 μLExtended LengthFilteredPack of 960
TFLR102-10-Q10 μLExtended LengthFilteredCase of 4800
TFLR102-10-QPK
TFLR102-10-Q으로도 사용됨
10 μLExtended LengthFilteredPack of 960
카탈로그 번호 TLR070RL-QPK
TLR070RL-Q으로도 사용됨
제품 가격(KRW)
78,000
Pack of 960
설명:
QSP 200, Non-Filtered
부피(미터법):
200 μL
팁 스타일:
Beveled
필터링됨:
Non-filtered
Unit Size:
Pack of 960
제품 가격(KRW)
78,000
Pack of 960
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
사양
클린 클레임DNase, RNase, DNA, and Pyrogen Free
설명QSP 200, Non-Filtered
길이(미터법)5.1cm
색상Natural
필터링됨Non-filtered
용도 (장비)Universal Fit
형식Refill
제품라인QSP
멸균Non-sterile
팁 스타일Beveled
부피(미터법)200 μL
Unit SizePack of 960