Search
Search

| 카탈로그 번호 | 설명 | 부피(미터법) | 팁 스타일 | 필터링됨 | 제품 사이즈 |
|---|---|---|---|---|---|
| TLR070RS-QPK TLR070RS-Q으로도 사용됨 | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Pack of 960 |
| TLR102-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Case of 10000 |
| TLR102-QPK TLR102-Q으로도 사용됨 | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Pack of 1000 |
| TLR102R-QPK TLR102R-Q으로도 사용됨 | QSP 10XL, Non-Filtered | 10 μL | Extended Length, Conical | Non-filtered | Pack of 960 |
| TLR102RS-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR102RS-QPK TLR102RS-Q으로도 사용됨 | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Pack of 960 |
| TLR102RL-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR102RL-QPK TLR102RL-Q으로도 사용됨 | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Pack of 960 |
| TLR102RLS-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR102RLS-QPK TLR102RLS-Q으로도 사용됨 | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered | Pack of 960 |
| TFLR113-20-Q | 20 μL | Beveled | Filtered | Case of 4800 | |
| TFLR113-20-QPK TFLR113-20-Q으로도 사용됨 | 20 μL | Beveled | Filtered | Pack of 960 | |
| TFLR113-100-Q | 100 μL | Beveled | Filtered | Case of 4800 | |
| TFLR113-100-QPK TFLR113-100-Q으로도 사용됨 | 100 μL | Beveled | Filtered | Pack of 960 | |
| TLR070-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Case of 10000 |
| TLR070-QPK TLR070-Q으로도 사용됨 | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Pack of 1000 |
| TLR070R-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Case of 4800 |
| TLR070R-QPK TLR070R-Q으로도 사용됨 | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Pack of 960 |
| TLR070RS-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Case of 4800 |
| TLR070RL-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Case of 4800 |
| TLR070RL-QPK TLR070RL-Q으로도 사용됨 | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Pack of 960 |
| TLR070RLS-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Case of 4800 |
| TLR070RLS-QPK TLR070RLS-Q으로도 사용됨 | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered | Pack of 960 |
| TLR106-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Case of 10000 |
| TLR106-QPK TLR106-Q으로도 사용됨 | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Pack of 1000 |
| TLR106R-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Case of 4800 |
| TLR106R-QPK TLR106R-Q으로도 사용됨 | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Pack of 960 |
| TLR106RS-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Case of 4800 |
| TLR106RS-QPK TLR106RS-Q으로도 사용됨 | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Pack of 960 |
| TLR106RL-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Case of 4800 |
| TLR106RL-QPK TLR106RL-Q으로도 사용됨 | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Pack of 960 |
| TLR106RLS-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Case of 4800 |
| TLR106RLS-QPK TLR106RLS-Q으로도 사용됨 | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered | Pack of 960 |
| TFLR140-200-Q | 200 μL | MicroPoint | Filtered | Case of 4800 | |
| TFLR140-200-QPK TFLR140-200-Q으로도 사용됨 | 200 μL | MicroPoint | Filtered | Pack of 960 | |
| TFLR140-300-Q | 300 μL | MicroPoint | Filtered | Case of 4800 | |
| TFLR140-300-QPK TFLR140-300-Q으로도 사용됨 | 300 μL | MicroPoint | Filtered | Pack of 960 | |
| TLR112NXL-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Case of 10000 |
| TLR112NXL-QPK TLR112NXL-Q으로도 사용됨 | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Pack of 1000 |
| TLR112NXLR-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR112NXLR-QPK TLR112NXLR-Q으로도 사용됨 | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Pack of 960 |
| TLR112NXLRS-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR112NXLRS-QPK TLR112NXLRS-Q으로도 사용됨 | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Pack of 960 |
| TLR112NXLRL-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR112NXLRL-QPK TLR112NXLRL-Q으로도 사용됨 | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Pack of 960 |
| TLR112NXRLS-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Case of 4800 |
| TLR112NXRLS-QPK TLR112NXRLS-Q으로도 사용됨 | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered | Pack of 960 |
| TFLR1121000-Q | 1000 μL | Extended Length | Filtered | Case of 4800 | |
| TFLR1121000-QPK TFLR1121000-Q으로도 사용됨 | 1000 μL | Extended Length | Filtered | Pack of 960 | |
| TFLR102-10-Q | 10 μL | Extended Length | Filtered | Case of 4800 | |
| TFLR102-10-QPK TFLR102-10-Q으로도 사용됨 | 10 μL | Extended Length | Filtered | Pack of 960 |
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.