RP6 Windshielded Swinging Bucket Rotor
Actual product may vary
RP6 Windshielded Swinging Bucket Rotor
RP6 Windshielded Swinging Bucket Rotor
Thermo Scientific™
RP6 Windshielded Swinging Bucket Rotor
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDescription
11178119Not ApplicableRP6 Windshield Rotor Lid (required)
111781351000 mLSealed Round Bucket
111781361000 mLUnsealed Round Buckets
11178138250 mLUnsealed Rectangular Buckets
11178140250 mLSealed Rectangular Buckets
11178216Not ApplicableSealing Lid for Rectangular Bucket
111782526 x 1000 mLRP6 Windshield Swinging Bucket Rotor
25144058Not ApplicableSealing Lid for Round Bucket
Catalog number 11178119
Price (MXN)
-
Capacity:
Not Applicable
Description:
RP6 Windshield Rotor Lid (required)
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
  • Capacity: 6 × 1000mL (6L) with Round bucket, 12 × 250mL (3L) with Rectangular bucket
  • Max. speed/RCF: 4250rpm/5,735 x g with Round bucket, 5,392 x g with Rectangular rotor
Ordering Information:
Order rotor body (11178252) and buckets and inserts to complete assembly. Order 11178216 for rectangular bucket sealing cap and 25144058 for round bucket sealing cap. Contact your Sales Representative for information.
Includes:
Rotor body only
Compatible with:
Thermo Scientific KR4i and Sorvall HT6 Centrifuges
Specifications
DescriptionRP6 Windshield Rotor Lid (required)
For Use With (Equipment)Thermo Scientific Sorvall HT6 and KR4i Large Capacity Centrifuges
Quantity1
BiocontainmentYes
CapacityNot Applicable
TypeBucket Lid
Unit SizeEach