SoftFit-L™ Non-Filtered Low Retention Pipette Tips in Racks with Lift-off Lid

SoftFit-L™ Non-Filtered Low Retention Pipette Tips in Racks with Lift-off Lid

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3721-0520 μLSterile
3722-0520 μLNon-sterile
3751-05200 μLSterile
3752-05200 μLNon-sterile
Catalog number 3721-05
Price (MXN)
-
Volume (Metric):
20 μL
Sterility:
Sterile
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description20L, Non-Filtered, Sterile, Rack
Length (English)1.95 in.
Length (Metric)4.86cm
Model20L
ColorNatural
Color CodeRed
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatRack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)20 μL
Unit SizeCase of 4800