Search
| Catalog Number | Thread Style | Packaging | Closure Color |
|---|---|---|---|
| 374516BLU-WP1D | Internal | 10 bags of 48 tubes | Blue |
| 374516RED-WP1D | Internal | Bulk, 10 bags of 48 tubes | Red |
| 374516ORG-WP1D | Internal | 10 bags of 48 | Orange |
| 374516YEL-WP1D | Internal | 10 bags of 48 | Yellow |
| 374516GRN-WP1D | Internal | 10 bags of 48 | Green |
| 374516PUR-WP1D | Internal | 10 bags of 48 | Purple |
Use the automation friendly Thermo Scientific™ Nunc™ Universal Tri-coded Tubes to improve the efficiency and throughput of your sample storage workflow. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D, 1D linear and human readable barcode. Tri-coded tubes come sterile and offered in both racked and bulk formats. The 2D screwtop tubes meet a range of manual and automated long term storage needs and can be used in combination with Thermo Scientific barcode readers, arrayers and capping equipment.
Scalable
Innovative Universal Cap Design
Secure Tracking
Stringent Quality Control