Search
| Catalog Number | Color | Sterility | Max. RCF |
|---|---|---|---|
| 508-GRD-O-Q | Orange | Non-sterile | 30,000 x g |
| 508-GRD-G-Q | Green | Non-sterile | 30,000 x g |
| 502-GRD-V-Q | Violet | Non-sterile | 30,000 x g |
| 509-GRD-MIXED | Non-sterile | ||
| 502-GRD-R-Q | Red | Non-sterile | 30,000 x g |
| 508-GRD-A-Q | Amber | Non-sterile | 30,000 x g |
| 502-GRD-Y-Q | Yellow | Non-sterile | 30,000 x g |
| 502-GRD-O-Q | Orange | Non-sterile | 30,000 x g |
| 509-GRD-G-Q | Green | Non-sterile | 30,000 x g |
| 509-GRD-Y-Q | Yellow | Non-sterile | 30,000 x g |
| 502-GRD-B-Q | Blue | Non-sterile | 30,000 x g |
| 502-GRDS-Q | Natural | Sterile | 30,000 x g |
| 509-GRD-R-Q | Red | Non-sterile | 30,000 x g |
| 508-GRD-R-Q | Red | Non-sterile | 30,000 x g |
| 508-GRD-B-Q | Blue | Non-sterile | 30,000 x g |
| 509-GRD-A-Q | Amber | Non-sterile | 30,000 x g |
| 502-GRD-A-Q | Amber | Non-sterile | 30,000 x g |
| 509-GRDS-Q | Natural | Sterile | 30,000 x g |
| 509-GRD-O-Q | Orange | Non-sterile | 30,000 x g |
| 508-GRD-V-Q | Violet | Non-sterile | 30,000 x g |
| 502-GRD-MIXED-Q | Assorted | Non-sterile | 30,000 x g |
| 508-GRDS-Q | Natural | Sterile | 30,000 x g |
| 508-GRD-Y-Q | Yellow | Non-sterile | 30,000 x g |
| 508-GRD-MIXED-Q | Assorted | Non-sterile | 30,000 x g |
| 509-GRD-MIXED-Q | Assorted | Non-sterile | 30,000 x g |
| 509-GRD-B-Q | Blue | Non-sterile | 30,000 x g |
| 502-GRD-G-Q | Green | Non-sterile | 30,000 x g |
| 509-GRD-V-Q | Violet | Non-sterile | 30,000 x g |
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube 0.6ml Graduated at 0.2, 0.4, 0.6ml, withstand up to 30,000 x g, frosted tube and cap and Amber color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.