Product Image
Thermo Scientific™
Thermo Scientific™ QSP™ Screw Cap Microcentrifuge Tube Caps
Thermo Scientific™ QSP™ Screw Cap Microcentrifuge Tube Caps with O-ring are designed to create a secure seal with the Thermo Scientific™ QSP™ micro tube providing Pure Protection for sample preparation, transfer and storage applications. Available in a rainbow of colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorAutoclavable
525-V-QVioletAutoclavable
525-A-QAmberAutoclavable
525-B-QBlueAutoclavable
525-G-QGreenAutoclavable
525-MIXED-QAssortedAutoclavable
525-O-QOrangeAutoclavable
525-QClearAutoclavable
525-R-QRedAutoclavable
525-Y-QYellowAutoclavable
TC-525-A-QAmberNot Autoclavable
TC-525-B-QBlueNot Autoclavable
TC-525-G-QGreenNot Autoclavable
TC-525-MIXED-QAssortedNot Autoclavable
TC-525-O-QOrangeNot Autoclavable
TC-525-QClearNot Autoclavable
TC-525-R-QRedNot Autoclavable
TC-525-V-QVioletNot Autoclavable
TC-525-Y-QYellowNot Autoclavable
Catalog number 525-V-Q
Price (MXN)
-
Color:
Violet
Autoclavable:
Autoclavable
Thermo Scientific™ QSP™ Screw Cap Microcentrifuge Tube Caps with O-rings, Amber, packaged bulk bag non-sterile with clean claims; RNase/DNase, DNA, pyrogen and endotoxin free.
The robust design and material selection are great for use with biologics, sample preparation, storage and transportation.
  • Screw Cap micro tube volume range of 0.5, 1.5 and 2.0ml and are ideal for sample preparation, storage and transportation
  • Conical bottom or self standing offered with knurls in ultra clear natural or UV protective amber color
  • Tether caps and standard non tether caps are available in a rainbow of color options
Specifications
Clean ClaimsRNase/DNase, DNA, pyrogen and endotoxin free
AutoclavableAutoclavable
ColorViolet
GraduatedYes
MaterialPolypropylene
SterilityNon-sterile
Unit SizeCase of 10