Search
Remove variability and achieve highly consistent results. High reproducibility of Thermo Scientific™ SOLAμ™ SPE plates deliver greater sample success at low elution volumes.
| Catalog Number | Includes |
|---|---|
| 60209-001RW | HRP 2 mg/1 mL 96 well removable plate |
| 60209-002RW | SCX 2 mg/1 mL 96 well removable plate |
| 60209-003RW | SAX 2 mg/1 mL 96 well removable plate |
| 60209-004RW | WCX 2 mg/1 mL 96 well removable plate |
| 60209-005RW | WAX 2 mg/1 mL 96 well removable plate |
Ideal for Bioanalytical and Clinical Research Laboratories
Bioanalytical laboratories continue to drive to higher efficacy drugs and long acting formulations which require lower quantification limits. There is also the desire to take advantage of the replacement, refinement and reduction policy, which is resulting in reduced animal model use. The growth of biopharmaceuticals also brings into consideration additional analytical issues such as solvation and non-specific binding.
Meet Demanding Sample Preparation Requirements
SOLAμ is the first micro-elution product to meet the requirements of the bioanalyst in combination with a robust, easy to use format.