nanoViper™ Capillary Kits for Low-Flow UHPLC Systems
Thermo Scientific™
nanoViper™ Capillary Kits for Low-Flow UHPLC Systems
Collect precise and reproducible low flow LC data with advanced nanoViper™ Fitting Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
6250.1030Tandem Workflow KitVanquish Neo UHPLC System
6720.0395nanoViper EASY-Spray Connection KitUltiMate 3000 RSLC Nano System
6720.0345nanoViper MS Connection KitLow-Flow UHPLC Systems
6250.1010Standard Application KitVanquish Neo UHPLC System
Catalog number 6250.1030
Price (MXN)
-
Description:
Tandem Workflow Kit
For Use With (Equipment):
Vanquish Neo UHPLC System
Avoid data-destructive dead volumes when connecting your low-flow UHPLC System. Ensure easy connections with the Thermo Scientific™ Standard Application Kit for Thermo Scientific™ Vanquish™ Neo UHPLC System for multiple nano-, capillary-, and micro-flow LC-MS workflows. Enhance your Vanquish Neo UHPLC system productivity with the tandem workflow kit. Use the Thermo Scientific™ nanoViper™ MS Connection Kits for Thermo Scientific™ EASY-Spray™ Columns for ideal column connections to a Thermo Scientific™ UltiMate™ 3000 RSLCnano System. Use the dedicated MS connection kit for all other connections of the UltiMate 3000 RSLCnano system to an MS inlet source. A single kit contains all the connectors and capillaries you need for tight and consistent connections.
See Viper and nanoViper resources
Experience the easy installation and robust performance of LC-MS connections with the nanoViper Fitting System kits for low-flow UHPLC Systems. Our nanoViper Fittings are designed to seal at the tip of the tubing to virtually eliminate dead volume and ensure easy connections in your low-flow UHPLC system.
Standard applications kits for the Vanquish Neo UHPLC system enable direct injection, trap-and-elute, and heated trap-and-elute LC-MS workflows. The tandem workflow kit extends Vanquish Neo system capabilities to include the tandem direct injection workflow. All kits facilitate dispersion-optimized MS front-end installation of the Thermo Scientific low-flow LC systems, including UltiMate 3000 RSLCnano systems, EASY-nLC 1200 systems, and Vanquish Neo UHPLC systems.
These kits feature:
  • Tailored kits for easy installation of MS front ends
  • Virtually zero-dead volume connections
  • Easy-to-install, reusable capillaries
  • Low carryover and simplified method transfer
  • Reliable and reproducible LC connections
Specifications
DescriptionTandem Workflow Kit
For Use With (Application)Tandem Direct Injection Workflow
IncludesCapillaries, Fittings, Standards, Columns (2)
Max. Pressure1500 bar
For Use With (Equipment)Vanquish Neo UHPLC System
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach