Search
Search
| Catalog Number | Type | For Use With (Equipment) |
|---|---|---|
| 7219-2134-003 | Tube Furnace Sleeve, 3 in. | STF54434C Tube Furnace |
| 59510 | Tube Furnace Adapter, 0.75 in. | HTF55122 Tube Furnace |
| 59511 | Tube Furnace Adapter, 1 in. | HTF55122 Tube Furnace |
| 59519 | Tube Furnace Blank Adapter | HTF55122 Tube Furnace |
| 59522 | Tube Furnace Adapter, 1.5 in | HTF55322, HTF55342, HTF55347 Furnace |
| 59523 | Tube Furnace Adapter, 2 in. | HTF55322, HTF55342, HTF55347 Furnace |
| 59524 | Tube Furnace Adapter, 2.5 in. | HTF55322, HTF55342, HTF55347 Furnace |
| 59525 | Tube Furnace Adapter, 3 in. | HTF55322, HTF55342, HTF55347 Furnace |
| 59529 | Tube Furnace Blank Adapter | HTF55322, HTF55342, HTF55347 Furnace |
| 59535 | Tube Furnace Adapter, 3 in. | HTF55667 Tube Furnace |
| 59536 | Tube Furnace Adapter, 4 in. | HTF55667 Tube Furnace |
| 59537 | Tube Furnace Adapter, 5 in. | HTF55667 Tube Furnace |
| 59541 | Tube Furnace Adapter, 1 in. | STF55346C Tube Furnace |
| 59543TA | Tube Furnace Adapter, 2 in. | STF55346C Tube Furnace |
| 59545 | Tube Furnace Adapter, 3 in. | STF55346C Tube Furnace |
| 59549 | Tube Furnace Blank Adapter | STF55346C Tube Furnace |
| 59555 | Tube Furnace Adapter, 3 in. | STF55666C Tube Furnace |
| 59556 | Tube Furnace Adapter, 4 in. | STF55666C Tube Furnace |
| 59557 | Tube Furnace Adapter, 5 in. | STF55666C Tube Furnace |
| 59558 | Tube Furnace Adapter, 6 in. | STF55666C Tube Furnace |
| 7219-2134-001 | Tube Furnace Sleeve, 1 in. | STF54434C Tube Furnace |
| 7219-2134-002 | Tube Furnace Sleeve, 2 in. | STF54434C Tube Furnace |
| 7219-2134-011 | Tube Furnace Sleeve, 3 in. | STF54454C Tube Furnace |
| 7219-2134-012 | Tube Furnace Sleeve, 2 in. | STF54454C Tube Furnace |
| 7219-2134-013 | Tube Furnace Sleeve, 1 in. | STF54454C Tube Furnace |
| 7219-2147-002 | Tube Furnace Adapter, 2 in. | STF54434C Tube Furnace |
| 7219-2147-003 | Tube Furnace Adapter, 3 in. | STF54434C Tube Furnace |
| 7219-2147-012 | Tube Vestibule, 2 in. | STF54454C Tube Furnace |
| 7219-2147-013 | Tube Vestibule, 1 in. | STF54454C Tube Furnace |