Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Actual product may vary
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Thermo Scientific™
Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeHoldsFor Use With (Equipment)
75004125Separatory Funnel ClampSeparatory FunnelSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30129Flask Clamp KitFlasksSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30150BIFlask Clamp10 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30151Flask Clamp25 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30152BIFlask Clamp50 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30153Flask Clamp125 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30154BIFlask Clamp250 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30155Flask Clamp300 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30156BIFlask Clamp500 mL Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30157BIFlask Clamp1 L Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30158Flask Clamp2 L Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30159Flask Clamp4 L Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30160Flask Clamp5 L Erlenmeyer FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30161Flask Clamp2500 mL Low-form Culture FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30162Flask Clamp2800 mL Fernbach FlaskSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
30175Microplate/Deep-Well Plate ClampMicroplate/Deep-Well PlateSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004101Vessel ClampSingle VesselSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004102Dual Height Adjustable ClampVesselSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004103Dual Height Adjustable ClampVesselSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004104Single Height Adjustable ClampVesselSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004106Media Bottle Clamp125 mL Square Media BottleSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004107Media Bottle Clamp250 mL Square Media BottleSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004108Media Bottle Clamp500 mL Square Media BottleSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
75004109Media Bottle Clamp1000 mL Square Media BottleSolaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
75004124Adhesive Mat applicatorVesselsSolaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
Catalog number 75004125
Price (MXN)
-
Product Type:
Separatory Funnel Clamp
Holds:
Separatory Funnel
For Use With (Equipment):
Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.
Specifications
Dimensions (L x W x H)403 x 159 x 174 mm platform
For Use With (Equipment)Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
HoldsSeparatory Funnel
Product TypeSeparatory Funnel Clamp
Unit SizeEach