Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
Actual product may vary
Thermo Scientific™
Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for Beckman™ automated liquid handling systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
935-262-0530 μL
915-262-0520 μL
938-262-0550 μL
917-262-05200 μL
Catalog number 935-262-05
Price (MXN)
-
Volume (Metric):
30 μL
Increase pipetting performance with Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
For Use With (Equipment)Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
FormatRack
SterilityNon-sterile
Tip StyleExtended Length
Volume (Metric)30 μL
Unit SizeEach