Hollow Cathode Lamps for iCE™ 3000 Series Atomic Absorption Spectrometers
Actual product may vary
Thermo Scientific™
Hollow Cathode Lamps for iCE™ 3000 Series Atomic Absorption Spectrometers
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series AAS system, we offer a huge range of pre-coded and un-coded Hollow Cathode Lamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeLamp
942339020511Hollow Cathode Lamps, UncodedAntimony Uncoded
942339020051Hollow Cathode Lamps, UncodedBoron Uncoded
942339020031Hollow Cathode Lamps, UncodedLithium Uncoded
942339020111Hollow Cathode Lamps, UncodedSodium Uncoded
942339020041Hollow Cathode Lamps, UncodedBeryllium Uncoded
942339020121Hollow Cathode Lamps, UncodedMagnesium Uncoded
94233902011VHollow Cathode Lamps, UncodedSodium (Na) Uncoded
942339020131Hollow Cathode Lamps, UncodedAluminum Uncoded
942339020141Hollow Cathode Lamps, UncodedSilicon Uncoded
94233902012VHollow Cathode Lamps, UncodedMagnesium (Mg) Uncoded
94233902013VHollow Cathode Lamps, UncodedAluminum (Al) Uncoded
942339020151Hollow Cathode Lamps, UncodedPhosphorus Uncoded
942339020201Hollow Cathode Lamps, UncodedCalcium Uncoded
942339020191Hollow Cathode Lamps, UncodedPotassium Uncoded
94233902019VHollow Cathode Lamps, UncodedPotassium (K) Uncoded
94233902020VHollow Cathode Lamps, UncodedCalcium (Ca) Uncoded
942339020231Hollow Cathode Lamps, UncodedVanadium Uncoded
942339020211Hollow Cathode Lamps, UncodedScandium Uncoded
942339020221Hollow Cathode Lamps, UncodedTitanium Uncoded
942339020241Hollow Cathode Lamps, UncodedChromium Uncoded
942339321241Hollow Cathode Lamps, UncodedCu/Ni Uncoded
942339321261Hollow Cathode Lamps, UncodedFe/Ni Uncoded
942339321231Hollow Cathode Lamps, UncodedCu/Mn Uncoded
942339321221Hollow Cathode Lamps, UncodedCu/Fe Uncoded
942339321301Hollow Cathode Lamps, UncodedCu/Mn/Ni Uncoded
942339321271Hollow Cathode Lamps, UncodedMn/Ni Uncoded
942339321291Hollow Cathode Lamps, UncodedCo/Mn/Ni Uncoded
942339321281Hollow Cathode Lamps, UncodedCo/Cu/Fe Uncoded
942339321311Hollow Cathode Lamps, UncodedCu/Mn/Fe Uncoded
942339321321Hollow Cathode Lamps, UncodedMn/Ni/Fe Uncoded
942339321341Hollow Cathode Lamps, UncodedCo/Cu/Mn/Ni Uncoded
942339321351Hollow Cathode Lamps, UncodedCo/Fe/Mn/Ni Uncoded
942339321331Hollow Cathode Lamps, UncodedCo/Cu/Fe/Mn Uncoded
942339321361Hollow Cathode Lamps, UncodedCu/Fe/Mn/Ni Uncoded
942339321371Hollow Cathode Lamps, UncodedCr/Mo Uncoded
942339321381Hollow Cathode Lamps, UncodedAl/Ti Uncoded
942339321411Hollow Cathode Lamps, UncodedAu/Ni Uncoded
942339321421Hollow Cathode Lamps, UncodedAu/Cu/Ni Uncoded
942339321401Hollow Cathode Lamps, UncodedAu/Cu Uncoded
942339321431Hollow Cathode Lamps, UncodedAg/Au Uncoded
942339321391Hollow Cathode Lamps, UncodedAl/Si/Ti Uncoded
942339321441Hollow Cathode Lamps, UncodedAg/Pt Uncoded
942339321461Hollow Cathode Lamps, UncodedAg/Au/Pt Uncoded
942339321451Hollow Cathode Lamps, UncodedAu/Pt Uncoded
942339321471Hollow Cathode Lamps, UncodedCa/Zn Uncoded
942339321511Hollow Cathode Lamps, UncodedFe/Zn Uncoded
942339321481Hollow Cathode Lamps, UncodedAg/Cr Uncoded
942339321501Hollow Cathode Lamps, UncodedAg/Cr/Cu Uncoded
942339321491Hollow Cathode Lamps, UncodedAg/Cu Uncoded
942339321521Hollow Cathode Lamps, UncodedMn/Zn Uncoded
942339321531Hollow Cathode Lamps, UncodedCu/Fe/Zn Uncoded
942339321541Hollow Cathode Lamps, UncodedCu/Mn/Zn Uncoded
942339321551Hollow Cathode Lamps, UncodedFe/Mn/Zn Uncoded
942339321581Hollow Cathode Lamps, UncodedCa/Sr Uncoded
942339321601Hollow Cathode Lamps, UncodedCa/Ba/Sr Uncoded
942339321561Hollow Cathode Lamps, UncodedCu/Fe/Mn/Zn Uncoded
942339321571Hollow Cathode Lamps, UncodedCa/Ba Uncoded
942339321591Hollow Cathode Lamps, UncodedBa/Sr Uncoded
942339321611Hollow Cathode Lamps, UncodedCd/Zn Uncoded
942339331011Hollow Cathode Lamp Data CodedCa/Mg Data Coded
942339331021Hollow Cathode Lamp Data CodedNa/K Data Coded
942339331031Hollow Cathode Lamp Data CodedCu/Zn Data Coded
942339331151Hollow Cathode Lamp Data CodedCr/Ni/Cu Data Coded
942339331141Hollow Cathode Lamp Data CodedCr/Mn Data Coded
942339331161Hollow Cathode Lamp Data CodedCr/Ni/Mn Data Coded
942339331171Hollow Cathode Lamp Data CodedCr/Ni/Cu/Mn/ Data Coded
942339331131Hollow Cathode Lamp Data CodedCr/Cu Data Coded
942339331191Hollow Cathode Lamp Data CodedCo/Fe Data Coded
942339331181Hollow Cathode Lamp Data CodedCo/Cu Data Coded
942339331201Hollow Cathode Lamp Data CodedCo/Mn Data Coded
942339331211Hollow Cathode Lamp Data CodedCo/Ni Data Coded
942339331221Hollow Cathode Lamp Data CodedCu/Fe Data Coded
942339331241Hollow Cathode Lamp Data CodedCu/Ni Data Coded
942339331231Hollow Cathode Lamp Data CodedCu/Mn Data Coded
942339331261Hollow Cathode Lamp Data CodedFe/Ni Data Coded
942339331251Hollow Cathode Lamp Data CodedFe/Mn Data Coded
942339331281Hollow Cathode Lamp Data CodedCo/Cu/Fe Data Coded
942339331291Hollow Cathode Lamp Data CodedCo/Mn/Ni Data Coded
942339331271Hollow Cathode Lamp Data CodedMn/Ni Data Coded
942339331301Hollow Cathode Lamp Data CodedCu/Mn/Ni Data Coded
942339331311Hollow Cathode Lamp Data CodedCu/Mn/Fe Data Coded
942339331331Hollow Cathode Lamp Data CodedCo/Cu/Fe/Mn Data Coded
942339331321Hollow Cathode Lamp Data CodedMn/Ni/Fe Data Coded
942339331351Hollow Cathode Lamp Data CodedCo/Fe/Mn/Ni Data Coded
942339331361Hollow Cathode Lamp Data CodedCu/Fe/Mn/Ni Data Coded
942339331341Hollow Cathode Lamp Data CodedCo/Cu/Mn/Ni Data Coded
942339331371Hollow Cathode Lamp Data CodedCr/Mo Data Coded
942339331401Hollow Cathode Lamp Data CodedAu/Cu Data Coded
942339331391Hollow Cathode Lamp Data CodedAl/Si/Ti Data Coded
942339331381Hollow Cathode Lamp Data CodedAl/Ti Data Coded
942339331431Hollow Cathode Lamp Data CodedAg/Au Data Coded
942339331411Hollow Cathode Lamp Data CodedAu/Ni Data Coded
942339331421Hollow Cathode Lamp Data CodedAu/Cu/Ni Data Coded
942339331441Hollow Cathode Lamp Data CodedAg/Pt Data Coded
942339331471Hollow Cathode Lamp Data CodedCa/Zn Data Coded
942339331451Hollow Cathode Lamp Data CodedAu/Pt Data Coded
942339331501Hollow Cathode Lamp Data CodedAg/Cr/Cu Data Coded
942339331461Hollow Cathode Lamp Data CodedAg/Au/Pt Data Coded
942339331481Hollow Cathode Lamp Data CodedAg/Cr Data Coded
942339331511Hollow Cathode Lamp Data CodedFe/Zn Data Coded
Catalog number 942339020511
Price (MXN)
-
Type:
Hollow Cathode Lamps, Uncoded
Lamp:
Antimony Uncoded
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific™ Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
Specifications
LampAntimony Uncoded
For Use With (Equipment)iCE 3000 Series Atomic Absorption Spectrometers
TypeHollow Cathode Lamps, Uncoded
Unit SizeEach