TSX Flex Series High Performance Laboratory Refrigerators
TSX Flex Series High Performance Laboratory Refrigerators
TSX Flex Series High Performance Laboratory Refrigerators
Thermo Scientific™

TSX Flex Series High Performance Laboratory Refrigerators

Green features
Thermo Scientific™ TSX Series high-performance, Innovative refrigeration solutions for healthcare and research needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Door StyleVoltage
TSX1-1205GV341 LGlass220 V
TSX1-1205SV341 LSolid220 V
TSX1-1205GA341 LGlass115 V
TSX1-1205SA341 LSolid115 V
TSX1-3605GV1031 LGlass220 V
TSX1-3605SV1031 LSolid220 V
TSX1-3605GA1031 LGlass115 V
TSX1-3605SA1031 LSolid115 V
Catalog number TSX1-1205GV
Price (MXN)
-
Capacity (Metric):
341 L
Door Style:
Glass
Voltage:
220 V
Thermo Scientific™ TSX HP Flex Laboratory Refrigerators are designed for laboratories requiring reliable storage of reagents, media, samples, kits, and other temperature-sensitive materials. Touchscreen controls, onboard data access, and adjustable interior configurations deliver an intuitive user experience while maintaining high-performance temperature control for everyday laboratory workflows

Reliable performance for everyday laboratory workflows

  • Designed for the storage of reagents, media, samples, kits, and other temperature-sensitive laboratory materials
  • Ideal for routine research, clinical, diagnostic, pharmaceutical, biotechnology, industrial, and academic laboratory environments
  • Designed to support convenient access and dependable storage of temperature-sensitive laboratory materials
  • Engineered to deliver dependable temperature performance for everyday laboratory applications

Designed to protect valuable samples

  • Designed to help maintain thermal stability and support efficient cold storage performance
  • Consistent temperature performance helps protect temperature-sensitive materials during routine laboratory use
  • Adjustable interior configurations help accommodate changing inventory and laboratory storage requirements
  • Designed to support dependable storage and efficient everyday laboratory workflows

Designed for confidence at the point of use

  • Modern touchscreen interface simplifies daily operation, configuration, and status review
  • Onboard data logging with USB download provides convenient access to temperature records for monitoring and documentation
  • Intuitive controls support faster training, easier setup, and greater confidence during daily operation
  • Designed to simplify routine monitoring without adding complexity to laboratory workflows

Compliance-ready cold storage

  • Designed to support qualification, validation, and audit-ready laboratory environments
  • cULus or CE listed, as appropriate by voltage and region
  • REACH, RoHS, Proposition 65, and WEEE compliant
  • North American 115V / 60Hz models are ENERGY STAR™ 2.0 certified, where applicable

Sustainable by design

  • Variable-speed compressor technology adapts cooling performance based on laboratory demand while maintaining stable temperatures
  • Designed to support compliance with EU F-Gas regulations and the U.S. EPA Significant New Alternatives Policy (SNAP)
  • Designed to help laboratories reduce energy consumption without compromising sample protection

Built for today's laboratory

  • Combines trusted TSX performance with improved usability, strong value, and product availability
  • Supports laboratories requiring dependable cold storage for regulated and routine environments
  • Designed to deliver the performance, compliance confidence, availability, and value laboratories expect
Specifications
Access SecurityKey Lock
Control TypeTouch Screen
Data OutputsUSB
DefrostAutomatic
Dimensions (D x W x H) Interior594 x 503 x 1353 mm
Dimensions (L x W x H) Exterior828 x 597 x 1649 mm
Exterior FinishPowder-coated Steel
Frequency50/60Hz
LightingLED
Warranty5 years parts and labor, plus an additional 5 years for compressor parts
Weight (English)317 lb.
Weight (Metric)144 kg
Capacity (English)12 cu.ft.
Capacity (Metric)341 L
Certifications/ComplianceCE, cULus, Prop 65, REACH, REACH
CompressorV-drive
Door StyleGlass
No. of Doors1
Plug TypeCEE 7/7
StyleUpright
Temperature Range2°C to 8°C
Voltage220 V
Unit SizeEach