Sodium Chloride (SQ), Qualigens
Qualigens

Sodium Chloride (SQ), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
50 kg
Packaging:
View all
Plastic Bottles
Catalog number Q27605
HSN 25010090
Price (INR)
320.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Specifications
Boiling Point1461°C
Melting Point801°C
CAS7647-14-5
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
View more

Detailed specifications PDF attached below