Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series’ V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
Product Line | TSX Series |
Description | TSX Series High-Performance Undercounter Refrigerator, Glass Door |
Temperature Range | 3°C to 7°C |
Certifications/Compliance | CE, REACH, RoHS, UL |
Capacity (English) | 5.5 cu. ft. |
Capacity (Metric) | 156 L |
Electrical Requirements | 100 - 240 V 50-60 Hz |
Voltage | 100/240 V |
Frequency | 50/60 Hz |
Amperage | 10 A Max / 3 A Steady State |
Plug Type | NEMA 5-15 P, More Global Types Available |
Dimensions (L x W x H) Exterior | 26 x 23.6 x 31.8 in. (66 x 60 x 81 cm) |
Dimensions (D x W x H) Interior | 20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4 cm) |
Weight (English) | 119 lb. |
Weight (Metric) | 53.98 kg |
Shipping Weight (English) | 155 lb. |
Shipping Weight (Metric) | 71 kg |
Display | Full color TFT-LCD; 4.3”; 480 x 272 |
Refrigerant | R744 |
Smart View Compatibility | Smart-Vue Compatible |
Depth (English) Interior | 20.5 in. |
Depth (Metric) Interior | 52 cm |
Height (English) Exterior | 31.8 in. |
Height (English) Interior | 23 in. |
Height (Metric) Exterior | 81 cm |
Height (Metric) Interior | 58.4 cm |
Width (English) Interior | 19.5 in. |
Width (Metric) Interior | 49.5 cm |
Defrost | Automatic |
Width (English) Exterior | 23.6 in. |
Width (Metric) Exterior | 60 cm |
Depth (English) Exterior | 26 in. |
Type | High-Performance Refrigerator |
Depth (Metric) Exterior | 66 cm |
Green Features | Energy efficient |
Door Style | Single Glass |
Monitoring Options | Alarm |
No. of Doors | 1 |
Shelves | 3 |
Style | Undercounter |
Unit Size | Each |
Product Line | TSX Series |
Description | TSX Series High-Performance Undercounter Refrigerator, Solid Door |
Temperature Range | 3°C to 7°C |
Certifications/Compliance | CE, REACH, RoHS, UL |
Capacity (English) | 5.5 cu. ft. |
Capacity (Metric) | 156 L |
Electrical Requirements | 100 - 240 V 50-60 Hz |
Voltage | 100/240 V |
Frequency | 50/60 Hz |
Amperage | 10 A Max / 3 A Steady State |
Plug Type | NEMA 5-15 P, More Global Types Available |
Dimensions (L x W x H) Exterior | 26 x 23.6 x 31.8 in. (66 x 60 x 81 cm) |
Dimensions (D x W x H) Interior | 20.5 x 19.5 x 23 in.(52 x 49.5 x 58.4 cm) |
Weight (English) | 119 lb. |
Weight (Metric) | 53.98 kg |
Shipping Weight (English) | 155 lb. |
Shipping Weight (Metric) | 71 kg |
Display | Full color TFT-LCD; 4.3”; 480 x 272 |
Refrigerant | R744 |
Smart View Compatibility | Smart-Vue Compatible |
Depth (English) Interior | 20.5 in. |
Depth (Metric) Interior | 52 cm |
Height (English) Exterior | 31.8 in. |
Height (English) Interior | 23 in. |
Height (Metric) Exterior | 79.5 cm |
Height (Metric) Interior | 58.4 cm |
Width (English) Interior | 19.5 in. |
Width (Metric) Interior | 49.5 cm |
Defrost | Automatic |
Width (English) Exterior | 23.6 in. |
Width (Metric) Exterior | 60 cm |
Depth (English) Exterior | 26 in. |
Type | High-Performance Refrigerator |
Depth (Metric) Exterior | 66 cm |
Green Features | Energy efficient |
Door Style | Single Solid |
Holds | 45 Boxes per Level (9 x 5 Pattern), 225 Boxes per Shelf (5 Levels per Shelf, 2 Shelves per Unit), +45 Boxes (1 Additional Level Above Top Shelf) |
Monitoring Options | Alarm |
No. of Doors | 1 |
Shelves | 3 |
Style | Undercounter |
Unit Size | Each |
Product Line | TSX Series |
Description | TSX Series Vaccine Storage Kit, Solid Door |
Temperature Range | 2°C to 8°C |
Certifications/Compliance | CE, REACH, RoHS, UL |
Capacity (English) | 5.5 cu. ft. |
Capacity (Metric) | 156 L |
Electrical Requirements | 100 - 240 V 50-60 Hz |
Voltage | 100/240 V |
Frequency | 50/60 Hz |
Amperage | 10 A Max / 3 A Steady State |
Plug Type | NEMA 5-15 P |
Dimensions (L x W x H) Exterior | 26 x 23.6 x 31.8 in. (66 x 60 x 81 cm) |
Dimensions (D x W x H) Interior | 20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4 cm) |
Weight (English) | 119 lb. |
Weight (Metric) | 53.98 kg |
Shipping Weight (English) | 155 lb. |
Shipping Weight (Metric) | 71 kg |
Display | Full color TFT-LCD; 4.3”; 480 x 272 |
Refrigerant | R744 |
Smart View Compatibility | Smart-Vue Compatible |
Includes | Refrigerator, wire basket drawer, datalogger |
Depth (English) Interior | 20.5 in. |
Depth (Metric) Interior | 52 cm |
Height (English) Exterior | 31.8 in. |
Height (English) Interior | 23 in. |
Height (Metric) Exterior | 81 cm |
Height (Metric) Interior | 58.4 cm |
Width (English) Interior | 19.5 in. |
Width (Metric) Interior | 49.5 cm |
Defrost | Automatic |
Width (English) Exterior | 23.6 in. |
Width (Metric) Exterior | 60 cm |
Depth (English) Exterior | 26 in. |
Type | High-Performance Refrigerator |
Depth (Metric) Exterior | 66 cm |
Door Style | Single Solid |
Monitoring Options | Alarm |
No. of Doors | 1 |
Shelves | 3 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Door Style | Electrical Requirements | Plug Type | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TSX505GA | Each | Single Glass | 100 - 240 V 50-60 Hz | NEMA 5-15 P, More Global Types Available | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSX505SA | Each | Single Solid | 100 - 240 V 50-60 Hz | NEMA 5-15 P, More Global Types Available | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSX505SAVK | Each | Single Solid | 100 - 240 V 50-60 Hz | NEMA 5-15 P | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|