Thermo Scientific Chemicals

Potassium hexacyanocobaltate(III), 90+%, Thermo Scientific Chemicals

Catalog number: 023126.18
50 g, Each
Thermo Scientific Chemicals

Potassium hexacyanocobaltate(III), 90+%, Thermo Scientific Chemicals

Catalog number: 023126.18
50 g, Each
Quantity
Catalog number: 023126.18
also known as 023126-18
Price (TWD)

Chemical Identifiers

CAS
13963-58-1
IUPAC Name
cobalt(3+) tripotassium hexakis(iminomethanide)
Molecular Formula
C6CoK3N6
InChI Key
LGRDAQPMSDIUQJ-UHFFFAOYSA-N
SMILES
[K+].[K+].[K+].[Co+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Assay (unspecified)
90.1% min

Description

It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text