Thermo Scientific Chemicals

Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals

Catalog number: 068107.18
50 g, Each
Thermo Scientific Chemicals

Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals

Catalog number: 068107.18
50 g, Each
Quantity
Catalog number: 068107.18
also known as 068107-18
Price (TWD)

Chemical Identifiers

CAS
3425-46-5
IUPAC Name
potassium cyanoselanide
Molecular Formula
CKNSe
InChI Key
KYEKHFSRAXRJBR-UHFFFAOYSA-M
SMILES
[K+].[Se-]C#N
Assay (unspecified)
98.5% min

Description

Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text