Thermo Scientific Chemicals

9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 153931000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 153931000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
642-31-9
IUPAC Name
anthracene-9-carbaldehyde
Molecular Formula
C15H10O
InChI Key
YMNKUHIVVMFOFO-UHFFFAOYSA-N
SMILES
O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
Appearance (Form)
Crystalline powder or needles
HPLC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Yellow to yellow-green to yellow-brown
Melting point
103°C to 107°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text