3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%

CAS: 22014-01-3 | C17H24O3 | 276.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10514.18
also known as A10514-18
Price (TWD)
-
Request A Quote
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS22014-01-3
IUPAC Name(2E)-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enoic acid
Molecular FormulaC17H24O3
InChI KeyCTYWXRDQWMRIIM-BQYQJAHWSA-N
SMILESCC(C)(C)C1=CC(C=CC(O)=O)=CC(=C1O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to orange
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
RUO – Research Use Only
C Daquino.;MC Foti. Coupling and fast decarboxylation of aryloxyl radicals of 4-hydroxycinnamic acids with formation of stable p-quinomethanes. Tetrahedron. 200662,(7) , 1536-1547.