Thermo Scientific Chemicals

2-Chloroethyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: A11863.14
25 g, Each
Thermo Scientific Chemicals

2-Chloroethyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: A11863.14
25 g, Each
Quantity
Catalog number: A11863.14
also known as A11863-14
Price (TWD)

Chemical Identifiers

CAS
80-41-1
IUPAC Name
2-chloroethyl 4-methylbenzene-1-sulfonate
Molecular Formula
C9H11ClO3S
InChI Key
ZXNMIUJDTOMBPV-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)OCCCl
Form
Liquid
Appearance (Color)
Clear or slightly turbid colourless to pale yellow
Assay (GC)
> 96.0%
Refractive Index
1.5265 - 1.5315 @20?C
Comment
may form a small amount of precipitate on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text