MagMAX™ Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit
MagMAX™ Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit
Applied Biosystems™

MagMAX™ Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit

The MagMAX Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit is a magnetic bead technology-based nucleic acid purification kit intended forRead more
Have Questions?
Catalog NumberQuantity
A52076192 reactions
Catalog number A52076
Price (TWD)
19,620.00
Online offer
Ends: 31-Dec-2025
21,800.00
Save 2,180.00 (10%)
Each
Add to cart
Quantity:
192 reactions
Price (TWD)
19,620.00
Online offer
Ends: 31-Dec-2025
21,800.00
Save 2,180.00 (10%)
Each
Add to cart
The MagMAX Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit is a magnetic bead technology-based nucleic acid purification kit intended for the isolation and purification of viral RNA and DNA from human saliva and nasopharyngeal/oropharyngeal swabs for thedetection of SARS-CoV-2 virus, adenovirus, and influenza virus by real-time PCR.

Features of the MagMAX Dx Prefilled Viral/Pathogen Nucleic Acid Isolation Kit include:
• Automation-ready script designed for KingFisher Flex system
• Fast procedure allows for 96 samples to be processed in <30 minutes on KingFisher Flex system
• Flexible protocol accommodates sample inputs from oropharyngeal/nasopharyngeal swabs and saliva
• Elution volume: 50 μL

Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.
For In Vitro Diagnostic Use.
Specifications
For Use With (Application)Real-Time PCR, Next Generation Sequencing
High-throughput CompatibilityHigh-throughput compatible, automated protocols
No. of Reactions192
Quantity192 reactions
Shipping ConditionAmbient
Sample TypeNasopharyngeal Swabs, Oropharyngeal Swab, Saliva
TargetRNA, DNA
Unit SizeEach