2,5-Diaminohydroquinone dihydrochloride, 97%
2,5-Diaminohydroquinone dihydrochloride, 97%
2,5-Diaminohydroquinone dihydrochloride, 97%
2,5-Diaminohydroquinone dihydrochloride, 97%
Thermo Scientific Chemicals

2,5-Diaminohydroquinone dihydrochloride, 97%

CAS: 24171-03-7 | C6H10Cl2N2O2 | 213.058 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number B21969.03
also known as B21969-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS24171-03-7
IUPAC Name2,5-diaminobenzene-1,4-diol
Molecular FormulaC6H8N2O2
InChI KeyRLXBOUUYEFOFSW-UHFFFAOYSA-N
SMILESNC1=CC(O)=C(N)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale grey to dark grey or purple to brown
FormPowder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. J. Szita and C. S. Marvel. Polymers derived from 2,5-diamino-p-benzoquinone-diimide and related compounds. Journal of Polymer Science Part A-1: Polymer Chemistry. 1969, 7 (11), 3203-3217.
  2. Z. Ahmad; F. Al Sagheer; A. Al Arbash; A.A.M. Ali. Synthesis and characterization of chemically cross-linked polyimide-siloxane hybrid films. Journal of Non-Crystalline Solids. 2009, 355 (8), 507-517.