Thermo Scientific Chemicals

1,4-Bis(2-hydroxyethyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: H55063.06
5 g, Each
Thermo Scientific Chemicals

1,4-Bis(2-hydroxyethyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: H55063.06
5 g, Each
Quantity
Catalog number: H55063.06
also known as H55063-06
Price (TWD)

Chemical Identifiers

CAS
122-96-3
IUPAC Name
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethan-1-ol
Molecular Formula
C8H18N2O2
InChI Key
VARKIGWTYBUWNT-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCO)CC1
Proton NMR
>97.5%

Description

It is applied for desulfurization, decarburization extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for desulfurization, decarburization extraction.

Solubility
Soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text