Methyl 5-bromo-2-methylbenzoate, 98%
Methyl 5-bromo-2-methylbenzoate, 98%
Methyl 5-bromo-2-methylbenzoate, 98%
Methyl 5-bromo-2-methylbenzoate, 98%
Thermo Scientific Chemicals

Methyl 5-bromo-2-methylbenzoate, 98%

CAS: 79669-50-4 | C9H9BrO2 | 229.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H61861.22
also known as H61861-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS79669-50-4
IUPAC Namemethyl 5-bromo-2-methylbenzoate
Molecular FormulaC9H9BrO2
InChI KeyFDCYLMYCHALQJR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(Br)=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
Bhaskar Reddy Kusuma.; Laura B. Peterson.; Huiping Zhao.; George Vielhauer.; Jeffrey Holzbeierlein.; Brian S. J. Blagg. Targeting the Heat Shock Protein 90 Dimer with Dimeric Inhibitors. J. Med. Chem. 2011, 54, (18), 6234-6253.