Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Fisher Chemical™

Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™

Have Questions?
Catalog NumberQuantityPackaging
S-P525-53
also known as S/P525/53
500 gPlastic powder jar
Catalog number S-P525-53
also known as S/P525/53
Price (TWD)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS7558-79-4
ChEBICHEBI:34683
ColorWhite
Formula Weight141.96g/mol
IUPAC Namedisodium hydrogen phosphate
View more
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous