Thermo Scientific Chemicals

Iron(III) oxalate hexahydrate, Thermo Scientific Chemicals

產品號碼: 031116.06
5 g, Each
Thermo Scientific Chemicals

Iron(III) oxalate hexahydrate, Thermo Scientific Chemicals

產品號碼: 031116.06
5 g, Each
Quantity
產品號碼: 031116.06
亦稱為 031116-06
價格 (TWD)

化學識別

CAS
166897-40-1
IUPAC Name
diiron(3+) hexahydrate trioxalate
Molecular Formula
C6H12Fe2O18
InChI Key
FWXIZVVTJVNNRX-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.[Fe+3].[Fe+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Appearance (Color)
Yellow-green
Assay (Titration)
> 97.0%
Form
powder

說明

Iron(III) oxalate hexahydrate is used as a catalyst for the reductive and oxidative degradation of iopamidol under UV and visible light irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text