Thermo Scientific Chemicals

Sodium 1-hexanesulfonate monohydrate, 99%, Thermo Scientific Chemicals

產品號碼: 032190.18
50 g, Each
Thermo Scientific Chemicals

Sodium 1-hexanesulfonate monohydrate, 99%, Thermo Scientific Chemicals

產品號碼: 032190.18
50 g, Each
Quantity
產品號碼: 032190.18
亦稱為 032190-18
價格 (TWD)

化學識別

CAS
207300-91-2
IUPAC Name
sodium hexane-1-sulfonate hydrate
Molecular Formula
C6H15NaO4S
InChI Key
CLCGFJYKZGFGSQ-UHFFFAOYSA-M
SMILES
O.[Na+].CCCCCCS([O-])(=O)=O
Optical Absorbance at
200 nm: ≤ 0.300 AU
Optical Absorbance at
210 nm: ≤0.100AU
Appearance (Color)
White
Form
powder
Assay (Titration)
≥98.5%

說明

Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text