Thermo Scientific Chemicals

DL-Homocysteinethiolactone hydrochloride, 99%

產品號碼: 120830250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Homocysteinethiolactone hydrochloride, 99%

產品號碼: 120830250
25 g, Glass bottle, Each
Quantity
Packaging
產品號碼: 120830250
價格 (TWD)

化學識別

CAS
6038-19-3
IUPAC Name
hydrogen 3-aminothiolan-2-one chloride
Molecular Formula
C4H8ClNOS
InChI Key
ZSEGSUBKDDEALH-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC1CCSC1=O
Titration with HClO4
>=98.5 %
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Infrared spectrum
Conforms

說明

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text