N-Boc-3-bromo-L-phenylalanine, 95%
N-Boc-3-bromo-L-phenylalanine, 95%
N-Boc-3-bromo-L-phenylalanine, 95%
N-Boc-3-bromo-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Boc-3-bromo-L-phenylalanine, 95%

CAS: 82278-73-7 | C14H18BrNO4 | 344.205 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H52024.03
亦稱為 H52024-03
價格 (TWD)
-
申請報價
Quantity:
1 g
Request bulk or custom format
規格
CAS82278-73-7
Chemical Name or MaterialN-Boc-3-bromo-L-phenylalanine
Formula Weight344.2
IUPAC Name(2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
InChI KeyFBUDYESOPLBQIR-NSHDSACASA-N
檢視更多
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ewa Szymanska; Darryl S.Pickering; Birgitte Nielsen; Tommy N.Johansen. 3-substituted phenylalanines as selective AMPA- and kainate receptor ligands. Bioorganic & Medicinal Chemistry. 2009, 17, (17),6390-6401