1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
Thermo Scientific Chemicals

1-Trimethylsilyl-1,4-pentadiyne, 98%

CAS: 71789-10-1 | C8H12Si | 136.27 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
產品號碼 H53436.06
亦稱為 H53436-06
價格 (TWD)
-
申請報價
Quantity:
5 g
Request bulk or custom format
化學識別
CAS71789-10-1
規格Specification Sheet規格表
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with strong oxidizing agents and ammonia.
RUO – Research Use Only

General References:

  1. Rodríguez, A. R.; Spur, B. W. First total synthesis of 7(S),17(S)-Resolvin D5, a potent anti-inflammatory docosanoid. Tetrahedron Lett. 2005, 46 (21), 3623-3627
  2. Zheng, X.; Herberich, G. E. Borabenzene Derivatives. 33. 3,5-Dimethylborabenzene 1,3,4,5-Tetramethylimidazol-2-ylidene: The First Carbene Adduct of a Borabenzene. Organometallics 2000, 19 (19), 3751-3753.