Product Image
Invitrogen™
pAd/PL-DEST™ Gateway™ Vector Kit
The pAd⁄PL-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ adenoviral expression vector, pAd⁄PL-DEST™ vector for easy recombination-based cloning and adenoviral-based,Read more
Have Questions?
Catalog NumberQuantity
V494206 μg
Catalog number V49420
Price (MXN)
-
Quantity:
6 μg
The pAd⁄PL-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ adenoviral expression vector, pAd⁄PL-DEST™ vector for easy recombination-based cloning and adenoviral-based, transient expression of a target gene in dividing and non-dividing mammalian cells. The vector allows generation of an adenovirus containing the target gene where expression is driven by a promoter of choice. Alternatively, the vector may also be used to express small RNA molecules from their appropriate promoters.
Advantages
• High efficiency and rapid recombination cloning
• Produces high titer adenoviral stocks
• Efficient delivery of the gene to dividing and non-dividing mammalian cells in vitro or in vivo
• Allows gene of interest to be controlled by a promoter of choice
• Produces replication-incompetent virus for enhanced biosafety of the system
• Amenable for use in high-throughput applications
Key Features
• Gateway™ Technology for efficient and rapid cloning
• Promoterless vector that allows gene of interest to be controlled by a promoter of choice
• Human Ad5 sequences (ΔE3) and Viral Inverted Terminal Repeats (ITRs) for packaging of the expression construct into virions
• Ampicillin selection marker
Kit includes
• pAd⁄PL-DEST™ Vector
• pAd⁄CMV⁄V5-GW⁄lacZ control plasmid
For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeAdenoviral
For Use With (Application)Clone your own Promoter, Viral Expression
Product TypeAdenovirus Expression Vector
Quantity6 μg
Selection Agent (Eukaryotic)None
VectorpDEST
Cloning MethodGateway
Product LineGateway, ViraPower
PromoterNone (Promoterless)
Protein TagUntagged
Unit Size6 µg
Contents & Storage
• pAd⁄PL-DEST™ Vector 150 ng⁄μl in TEBuffer, pH 8.0. 40 μl (Store at -20°C)
• pAd⁄CMV⁄V5-GW⁄lacZ control plasmid 1 μg⁄μl in TE Buffer, pH 8.0. 10 μl (Store at -20°C)
Frequently asked questions (FAQs)
Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?
In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.
Do you have a recommended single-step protocol for BP/LR recombination?
Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.
How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?
We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.
Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?
We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.
Do you offer Gateway vectors for expression in plants?
We do not offer any Gateway vectors for expression in plants.
Citations & References (1)
Citations & References
Abstract
Pulmonary interleukin-23 gene delivery increases local T-cell immunity and controls growth of Mycobacterium tuberculosis in the lungs.
Authors:Happel KI, Lockhart EA, Mason CM, Porretta E, Keoshkerian E, Odden AR, Nelson S, Ramsay AJ,
Journal:Infect Immun
PubMed ID:16113296
Interleukin-23 (IL-23) is a heterodimeric cytokine that shares IL-12 p40 but contains a unique p19 subunit similar to IL-12 p35. Previous studies indicate a greater importance for intact IL-12/23 p40 expression than IL-12 p35 for immunity against Mycobacterium tuberculosis, suggesting a role for IL-23 in host defense. The effects of ... More