Gene Transcripts

Gene Symbol
Cox5a
Entrez Gene ID
12858
Gene Name
cytochrome c oxidase subunit Va
Gene Aliases
AA959768, CcOX
Chromosome Location
Chr. 9: 57521232 - 57532427 on Build GRCm38
UniGene ID
Mm.273403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007747.2 NP_031773.2 4 452
GenBank mRNA AK018723.1 - 4 387
BC003831.1 - 3 1578
BC034302.1 - 4 405 5372849
X15963.1 - 4 376

View Full Product Details