Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178061.5 | NP_835162.1 | 2 | 746 | |
GenBank mRNA | AK039841.1 | - | 2 | 592 | |
AK087285.1 | - | 2 | 747 | ||
AK155217.1 | - | 2 | 747 | ||
AK170973.1 | - | 2 | 740 | ||
BC020028.1 | - | 2 | 728 | 4007373 |