Gene Transcripts

Gene Symbol
Cyp3a44
Entrez Gene ID
337924
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 44
Gene Aliases
Chromosome Location
Chr. 5: 145773983 - 145805891 on Build GRCm38
UniGene ID
Mm.425301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177380.3 NP_796354.1 7 708
GenBank mRNA AB039380.1 - 7 708
BC125522.1 - 7 670 40130037

View Full Product Details