Gene Transcripts

Gene Symbol
Cxcl12
Entrez Gene ID
20315
Gene Name
chemokine (C-X-C motif) ligand 12
Gene Aliases
Pbsf, Scyb12, Sdf1, Tlsf, Tpar1
Chromosome Location
Chr. 6: 117168535 - 117181368 on Build GRCm38
UniGene ID
Mm.303231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012477.2 NP_001012495.1 2 264
NM_013655.4 NP_038683.1 2 264
NM_021704.3 NP_068350.1 2 264
GenBank mRNA AK045092.1 - 2 223
AK075596.1 - 2 223
AK157553.1 - 2 225
BB610070.1 - 2 220
BB840704.1 - 2 223
BC006640.1 - 2 212 3483088
BC020297.1 - 2 212
BC029335.1 - 2 212
BC046827.1 - 2 214
CT010389.1 - 2 143
D21072.1 - 2 201
D43804.1 - 2 221
D43805.1 - 2 221
L12029.1 - 2 224
L12030.1 - 2 219
S74318.1 - 2 209

View Full Product Details