Gene Transcripts
- Gene Symbol
- Cxcl12
- Entrez Gene ID
- 20315
- Gene Name
- chemokine (C-X-C motif) ligand 12
- Gene Aliases
- Pbsf, Scyb12, Sdf1, Tlsf, Tpar1
- Chromosome Location
- Chr. 6: 117168535 - 117181368 on Build GRCm38
- UniGene ID
- Mm.303231
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012477.2 |
NP_001012495.1
|
2, 3 |
303 |
|
NM_013655.4 |
NP_038683.1
|
2, 3 |
303 |
|
NM_021704.3 |
NP_068350.1
|
2, 3 |
303 |
|
GenBank mRNA |
AK045092.1 |
- |
2, 3 |
262 |
|
AK075596.1 |
- |
2, 3 |
262 |
|
AK157553.1 |
- |
2, 3 |
264 |
|
BB610070.1 |
- |
2, 3 |
259 |
|
BB840704.1 |
- |
2, 3 |
262 |
|
BC006640.1 |
- |
2, 3 |
251 |
3483088 |
BC020297.1 |
- |
2, 3 |
251 |
|
BC029335.1 |
- |
2, 3 |
251 |
|
BC046827.1 |
- |
2, 3 |
253 |
|
CT010389.1 |
- |
2, 3 |
182 |
|
D21072.1 |
- |
2, 3 |
240 |
|
D43804.1 |
- |
2, 3 |
260 |
|
D43805.1 |
- |
2, 3 |
260 |
|
L12029.1 |
- |
2, 3 |
263 |
|
L12030.1 |
- |
2, 3 |
258 |
|
S74318.1 |
- |
2, 3 |
248 |
|
View Full Product Details