Gene Transcripts

Gene Symbol
SCYL2
Entrez Gene ID
55681
Gene Name
SCY1 like pseudokinase 2
Gene Aliases
CVAK104
Chromosome Location
Chr. 12: 100267137 - 100341724 on Build GRCh38
UniGene ID
Hs.506481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317784.1 NP_001304713.1 7 1187
NM_017988.5 NP_060458.3 7 1339
XM_017019618.1 XP_016875107.1 7 1187
XM_017019619.1 XP_016875108.1 7 1339
XM_017019620.1 XP_016875109.1 8 1326
GenBank mRNA AB037781.1 - 4 493
AK000936.1 - 6 945
AK024274.1 - 3 348
AK027551.1 - 3 405
AK125091.1 - 7 1324
AK292980.1 - 7 1113
BC063798.1 - 7 952 6473586

View Full Product Details