Gene Transcripts

Gene Symbol
Rln3
Entrez Gene ID
212108
Gene Name
relaxin 3
Gene Aliases
M3, insl7
Chromosome Location
Chr. 8: 84043067 - 84044979 on Build GRCm38
UniGene ID
Mm.261583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173184.1 NP_775276.1 2 433
GenBank mRNA AB076565.1 - 2 433

View Full Product Details