Gene Transcripts

Gene Symbol
Lyl1
Entrez Gene ID
17095
Gene Name
lymphoblastomic leukemia 1
Gene Aliases
Lyl-1, bHLHa18
Chromosome Location
Chr. 8: 84701422 - 84704716 on Build GRCm38
UniGene ID
Mm.4925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008535.2 NP_032561.2 2 744
GenBank mRNA AK076114.1 - 2 398
BC005736.1 - 2 364 3598911
CT010167.1 - 1 189
X57687.1 - 2 654

View Full Product Details