Gene Transcripts
- Gene Symbol
- IFRD1
- Entrez Gene ID
- 3475
- Gene Name
- interferon related developmental regulator 1
- Gene Aliases
- PC4, TIS7
- Chromosome Location
- Chr. 7: 112423144 - 112477203 on Build GRCh38
- UniGene ID
- Hs.744747
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007245.2 |
NP_001007246.1
|
12, 13 |
1743 |
|
NM_001197079.1 |
NP_001184008.1
|
11, 12 |
1633 |
|
NM_001197080.1 |
NP_001184009.1
|
11, 12 |
1468 |
|
NM_001550.3 |
NP_001541.2
|
11, 12 |
1534 |
|
XM_011516142.2 |
XP_011514444.1
|
12, 13 |
1544 |
|
XM_011516143.2 |
XP_011514445.1
|
11, 12 |
1729 |
|
XM_017012151.1 |
XP_016867640.1
|
11, 12 |
1781 |
|
GenBank mRNA |
AK298894.1 |
- |
11, 12 |
1274 |
|
AK308066.1 |
- |
12, 13 |
1718 |
|
AK313211.1 |
- |
12, 13 |
1743 |
|
AK315967.1 |
- |
11, 12 |
1466 |
|
BC001272.1 |
- |
11, 12 |
1487 |
3458309 |
BI915700.1 |
- |
1, 2 |
33 |
|
BT019889.1 |
- |
11, 12 |
1273 |
|
BT019890.1 |
- |
11, 12 |
1273 |
|
BX648799.1 |
- |
3, 4 |
971 |
|
View Full Product Details