Gene Transcripts

Gene Symbol
Ctla2a
Entrez Gene ID
13024
Gene Name
cytotoxic T lymphocyte-associated protein 2 alpha
Gene Aliases
Ctla-2a
Chromosome Location
Chr. 13: 60934155 - 60936625 on Build GRCm38
UniGene ID
Mm.482089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145799.1 NP_001139271.1 2, 3 212
NM_007796.2 NP_031822.2 2, 3 250
GenBank mRNA AA451288.1 - 2, 3 212
AK005471.1 - 2, 3 187
AK018801.1 - 2, 3 205
AK132290.1 - 2, 3 183
AK167433.1 - 2, 3 183
AK167664.1 - 2, 3 184
AY034577.1 - 2, 3 159
BC028437.1 - 2, 3 176 1244311
X15591.1 - 2, 3 159

View Full Product Details