Gene Transcripts

Gene Symbol
Klk1b16
Entrez Gene ID
16615
Gene Name
kallikrein 1-related peptidase b16
Gene Aliases
Klk16, mGk-16
Chromosome Location
Chr. 7: 44136767 - 44141604 on Build GRCm38
UniGene ID
Mm.440887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008454.2 NP_032480.1 4 1076
GenBank mRNA BC012237.1 - 4 547
J03877.1 - 4 1077

View Full Product Details