Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025417.2 | NP_079693.1 | 4, 5 | 210 | |
GenBank mRNA | AK004174.1 | - | 4, 5 | 211 | |
AK009802.1 | - | 4, 5 | 209 | ||
AK172194.1 | - | 3, 4 | 1322 | ||
AK188693.1 | - | 3, 4 | 178 | ||
BC033438.1 | - | 3, 4 | 196 | 3995599 | |
BC049222.1 | - | 4, 5 | 209 | 2649359 | |
BC055798.1 | - | 3, 4 | 1301 |