Gene Transcripts
- Gene Symbol
- Hopx
- Entrez Gene ID
- 74318
- Gene Name
- HOP homeobox
- Gene Aliases
- 1110018K11Rik, 1200015P04Rik, 2300002F06Rik, AI848177, AW490897, Cameo, Hdop, Hod, Hop, Ob1, Obl, Toto
- Chromosome Location
- Chr. 5: 77086986 - 77115123 on Build GRCm38
- UniGene ID
- Mm.181852
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159900.1 |
NP_001153372.1
|
2, 3 |
276 |
|
NM_001159901.1 |
NP_001153373.1
|
1, 2 |
404 |
|
NM_175606.3 |
NP_783199.1
|
2, 3 |
436 |
|
GenBank mRNA |
AF492703.1 |
- |
2, 3 |
436 |
|
AF492704.1 |
- |
2, 3 |
264 |
|
AF534182.1 |
- |
2, 3 |
481 |
|
AF536202.1 |
- |
1, 2 |
173 |
|
AI604115.1 |
- |
1, 2 |
390 |
|
AK003784.1 |
- |
2, 3 |
265 |
|
AK008297.1 |
- |
2, 3 |
243 |
|
AK008439.1 |
- |
2, 3 |
243 |
|
AK009007.1 |
- |
2, 3 |
193 |
|
BC024546.1 |
- |
2, 3 |
260 |
4989797 |
View Full Product Details