Gene Transcripts
- Gene Symbol
- ASGR1
- Entrez Gene ID
- 432
- Gene Name
- asialoglycoprotein receptor 1
- Gene Aliases
- ASGPR, ASGPR1, CLEC4H1, HL-1
- Chromosome Location
- Chr. 17: 7173431 - 7179564 on Build GRCh38
- UniGene ID
- Hs.12056
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197216.2 |
NP_001184145.1
|
4, 5 |
633 |
|
NM_001671.4 |
NP_001662.1
|
5, 6 |
750 |
|
XM_011523861.2 |
XP_011522163.1
|
5, 6 |
657 |
|
GenBank mRNA |
AB070933.1 |
- |
|
529 |
|
AK308375.1 |
- |
5, 6 |
556 |
|
AK308677.1 |
- |
3, 4 |
381 |
|
BC032130.2 |
- |
5, 6 |
427 |
5087444 |
BQ653541.1 |
- |
5, 6 |
760 |
|
BT019873.1 |
- |
4, 5 |
350 |
|
CR542052.1 |
- |
4, 5 |
350 |
|
FJ410292.1 |
- |
3, 4 |
233 |
|
M10058.1 |
- |
5, 6 |
522 |
|
View Full Product Details