Gene Transcripts

Gene Symbol
ICK
Entrez Gene ID
22858
Gene Name
intestinal cell (MAK-like) kinase
Gene Aliases
ECO, LCK2, MRK
Chromosome Location
Chr. 6: 53001299 - 53061864 on Build GRCh38
UniGene ID
Hs.417022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014920.3 NP_055735.1 4 636
NM_016513.4 NP_057597.2 5 747
XM_011514419.1 XP_011512721.1 4 636
XM_011514420.2 XP_011512722.1 5 778
XM_011514421.1 XP_011512723.1 5 717
XM_017010485.1 XP_016865974.1 5 1010
XM_017010486.1 XP_016865975.1 6 889
XM_017010487.1 XP_016865976.1 4 555
XM_017010488.1 XP_016865977.1 5 1437
XM_017010489.1 XP_016865978.1 6 952
XM_017010490.1 XP_016865979.1 5 822
XM_017010491.1 XP_016865980.1 5 783
XM_017010492.1 XP_016865981.1 4 827
GenBank mRNA AB023153.1 - 4 523
AF152469.1 - 4 604
AF225919.1 - 4 466
AK074892.1 - 5 717
BC035807.1 - 4 668
BC136420.1 - 3 297 9020408
BC136421.1 - 3 297 9020409
BC152464.1 - 4 523 8862600

View Full Product Details